Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TSYVAESSTA EDALSIRSEM IQRKGSTFRP HDSFPKSGKS GRRRRERRST VLGLPQHVQK ELGLRNEREA PGTPRAPGAR DAVRIPTVD
Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 90%.
KIAA1522 gene ontology annotations related to this gene include cell differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: NHS-like protein 3
Gene Aliases: KIAA1522; NHSL3
UniProt ID: (Human) Q9P206
Entrez Gene ID: (Human) 57648
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support