Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDEEVSDPTS AAAQLRQLRD HLPPPSSASP SRALPEPQEA QKLAAERARA PVVPYGVDLH YWGQELPTAG KIVKSDSQHR FWKPSEVEEE VVNADISEML RSRHITFAGK FEPVQHWCRA PRPDGRLCER QDRLKCPFHG KIVPRDDEGR PLDPEDRARE QRRQLQKQER PEWQDPELMR DVEAATGQDL GSSRYSGKGR GKKRRYPSLT NLKAQADTAR ARIGRKVFAK AAVRRVVAAM NRMDQKKHEK FSNQFNYALN
The protein encoded by KIAA1530 appears to be involved in ubiquitination and dephosphorylation of RNA polymerase II subunits that stall after UV irradiation. The encoded protein interacts with several members of the nucleotide excision repair complex, and is thought to be involved in the transcription-coupled nucleotide excision repair (TC-NER) pathway to help remove lesions in the DNA that block transcription. Defects in KIAA1530 can cause UV-sensitive syndrome 3. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: UV-stimulated scaffold protein A
Gene Aliases: KIAA1530; UVSS3; UVSSA
UniProt ID: (Human) Q2YD98
Entrez Gene ID: (Human) 57654
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support