Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: NKTTRQASLR KKIREHDVSK AHGKIQDLLK ESTNDSICNL VHKQNNKNID ATVKVFNTVY SLVKHNRPLS DIEGARELQE KNGEVNCLNT RYSATRI
Highest antigen sequence identity to the following orthologs: Mouse - 24%, Rat - 24%.
E3 SUMO-protein ligase; facilitates UBE2I/UBC9-mediated SUMO2 modification of target proteins (PubMed:26524493).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 SUMO-protein ligase KIAA1586; E3 SUMO-protein transferase KIAA1586
Gene Aliases: KIAA1586
UniProt ID: (Human) Q9HCI6
Entrez Gene ID: (Human) 57691
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support