Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKSSVAQIKP SSGHDRRENL NSYQRNSSPE DRYEEQERSP RDRDYFDYSR SDYEHSRRGR SYDSSMESRN RDREKRRERE RDTDRKRSRK SPSPGRRNPE TSVTQSSSAQ DEPATKKKKD ELDPLLTRTG GAYIPPAKLR MMQEQITDKN SLAYQRMSWE ALKKSINGLI NKVNISNISI IIQELLQENI VRGRGLLSRS VLQAQSASPI FTHVYAALVA IINSKFPQIG ELILKRLILN FRKGYRRNDK QLCLTASKFV AHLINQNVAH EVLCLEMLTL LLERPTDDSV EVAIGFLKEC GLKLTQVSPR GINAIFERLR NILHESEIDK RVQYMIEVMF AVRKDGFKDH PIILEGLDLV EEDDQFTHML PLEDDYNPED VLNVFKMDPN FMENEEKYKA IKKEILDEGD TDSNTDQDAG SSEEDEEEEE EEGEEDEEGQ KVTIHDKTEI NLVSFRRTIY LAIQSSLDFE ECAHKLLKME FPESQTKELC NMILDCCAQQ RTYEKFFGLL AGRFCMLKKE YMESFEGIFK EQYDTIHRLE TNKLRNVAKM FAHLLYTDSL PWSVLECIKL SEETTTSSSR IFVKIFFQEL CEYMGLPKLN ARLKDETLQP FFEGLLPRDN PRNTRFAINF FTSIGLGGLT DELREHLKNT PKVIVAQKPD VEQNKSSPSS SSSASSSSES DSSDSDSDSS DSSSESSSEE SDSSSISSHS SASANDVRKK GHGKTRSKEV DKLIRNQQTN DRKQKERRQE HGHQETRTER ERRSEKHRDQ NSRGSNWRDP ITKYTSDKDV PSERNNYSRV ANDRDQEMHI DLENKHGDPK KKRGERRNSF SENEKHTHRI KDSENFRRKD RSKSKEMNRK HSGSRSDEDR YQNGAERRWE KSSRYSEQSR ESKKNQDRRR EKSPAKQK
Required for pre-mRNA splicing as component of the spliceosome (PubMed:12226669, PubMed:11991638, PubMed:22961380, PubMed:28502770, PubMed:28076346, PubMed:29360106, PubMed:29301961). Promotes exon-junction complex (EJC) assembly (PubMed:22959432, PubMed:22961380). Hinders EIF4A3 from non-specifically binding RNA and escorts it to the splicing machinery to promote EJC assembly on mature mRNAs. Through its role in EJC assembly, required for nonsense-mediated mRNA decay. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CWC22 spliceosome-associated protein homolog; fSAPb; functional spliceosome-associated protein b; Nucampholin homolog; Pre-mRNA-splicing factor CWC22 homolog
Gene Aliases: CWC22; EIF4GL; fSAPb; KIAA1604; NCM
UniProt ID: (Human) Q9HCG8
Entrez Gene ID: (Human) 57703
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support