Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive test controls include: HeLa, MCF7, NIH/3T3.
Immunogen sequence: MDPQRSPLLE VKGNIELKRP LIKAPSQLPL SGSRLKRRPD QMEDGLEPEK KRTRGLGATT KITTSHPRVP SLTTVPQTQG QTTAQKVSKK TGPRCSTAIA
Minus end-directed microtubule-dependent motor required for bipolar spindle formation (PubMed:15843429). May contribute to movement of early endocytic vesicles. Regulates cilium formation and structure. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kifc1; kinesin family c-terminal 5A; kinesin motor protein KIFC4; kinesin-like 2; kinesin-like 2a; Kinesin-like protein 2; Kinesin-like protein KIFC1; Kinesin-related protein HSET; kinesin-related protein KIFC5A
Gene Aliases: HSET; 100042970; Gm4137; KIFC1; Kifc4; Kifc5a; Kifc5b; KNSL2; Knsl2a; Tctex-7; Tctex-7A; Tctex7; Tctex7a
UniProt ID: (Human) Q9BW19, (Mouse) Q9QWT9
Entrez Gene ID: (Human) 3833, (Mouse) 100502766
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support