Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody is stable for 24 months.
Positive Control: HeLa cells. Cellular Localization: Nucleus.
Specificity Comments: Kr ppel-like factors (KLFs) comprise a family of evolutionarily conserved zinc finger-containing transcription factors with diverse regulatory functions in cell growth, proliferation, differentiation and embryogenesis. Individual members of the Sp1-like/KLF family can function either as activators or repressors, depending on which promoter they bind and which co-regulators they interact with. KLF17 (Kr ppel-like factor 17), whose alternative names include ZNF393 (zinc finger protein 393) or zfp393, is a 389 amino acid nuclear protein belonging to the Sp1 C2H2-type zinc-finger protein family. Expressed in testis and ovary, KLF17 may function as a germ cell-specific transcription factor involved in oocyte development and spermatid differentiation. Containing three C2H2-type zinc fingers which bind G/C-rich sites, KLF17 activates transcription from CACCC-box elements.
KLF17 is a transcription repressor that binds to the promoter of target genes and prevents their expression. Acts as a negative regulator of epithelial-mesenchymal transition and metastasis in breast cancer. Specifically binds the 5'-CACCC-3' sequence in the promoter of ID1, a key metastasis regulator in breast cancer, and repress its expression. May be a germ cell-specific transcription factor that plays important roles in spermatid differentiation and oocyte development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ40160; Krueppel-like factor 17; Kruppel-like factor 17; novel zinc-finger protein; Zinc finger protein 393; ZLF393
Gene Aliases: KLF17; Zfp393; ZNF393
UniProt ID: (Human) Q5JT82
Entrez Gene ID: (Human) 128209
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support