Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: FHEVLCKPQ FHLLGSPPQA PGDVSLKQRL REARMTGTPV LVALGDFLGG PLAPHPYQGE PPSMLRYEEM TERWFPLANN LPPDLVNVRG YGSAILDNYL FIVGGYRITS QEISAAHSYN PSTNEWLQVA SMNQKRSNFK LVAVNSKLYA IGGQAVSNVE CYNPEQDAWN FVAPLPNPLA EFSACECKGK IYVIGGYTTR DRNMNILQYC PSSDMWTLFE TCDVHIRKQQ MVSVEETIYI VGGCLHELGP NRRSSQSEDM LTVQSYNTVT RQWLYLKENT SKSGLNLTCA LHNDGIYIMS RDVTLSTSLE HRVFLKYNIF SDSWEAFRRF PAFGHNLLVS SLYLPNKAET (157-505 aa encoded by BC137116)
Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex required for mitotic progression and cytokinesis. The BCR(KLHL42) E3 ubiquitin ligase complex mediates the ubiquitination and subsequent degradation of KATNA1. Involved in microtubule dynamics throughout mitosis. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ctb9; Cullin-3-binding protein 9; kelch domain containing 5; Kelch domain-containing protein 5; kelch-like family member 42; kelch-like family, member 42; Kelch-like protein 42
Gene Aliases: A330079N23Rik; AI450355; C230080I20Rik; Ctb9; KIAA1340; KLHDC5; KLHL42; mKIAA1340
UniProt ID: (Human) Q9P2K6, (Mouse) Q8BFQ9
Entrez Gene ID: (Human) 57542, (Mouse) 232539
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support