Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
KLHDC9 gene ontology annotations related to this gene include cyclin binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Kelch domain-containing protein 9; Kelch/ankyrin repeat-containing cyclin A1-interacting protein
Gene Aliases: 1190002J23Rik; AA087357; ESTM31; KARCA1; KLHDC9; RGD1561025
UniProt ID: (Human) Q8NEP7, (Mouse) Q3USL1
Entrez Gene ID: (Human) 126823, (Mouse) 68874, (Rat) 360878
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support