Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSRSGDRTS TFDPSHSDNL LHGLNLLWRK QLFCDVTLTA QGQQFHCHKA VLASCSQYFR SLFSSHPPLG GGVGGQDGLG APKDQQQPPQ QQPSQQQQPP PQEEPGTPSS SPDDKLLTSP RAINNLVLQG CSSIGLRLVL EYLYTANVTL SLDTVEEVLS VSKILHIPQV TKLCVQFLND QISVQNYKQV CKIAALHGLE ETKKLANKYL VEDVLLLNFE EMRALLDSLP PPVESELALF QMSVLWLEHD RETRMQYAPD LMKRLRFALI PAPELVERVQ SVDFMRTDPV CQKLLLDAMN YHLMPFRQHC RQSLASRIRS NKKMLLLVGG LPPGPDRLPS NLVQYYDDEK KTWKILTITG PHPISGKQES ASSPEFRRRK TREFSNNTM (1-388 aa encoded by BC021267 )
KLHL14 is a member of the KLHL family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kelch-like 14; kelch-like family member 14; Kelch-like protein 14; Printor; Protein interactor of Torsin-1A; protein interactor of torsinA
Gene Aliases: 6330403N15Rik; AI785250; KIAA1384; KLHL14; printor; RGD1566178
UniProt ID: (Human) Q9P2G3, (Mouse) Q69ZK5
Entrez Gene ID: (Human) 57565, (Rat) 364823, (Mouse) 225266
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support