Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Predicted reactivity by homology: Mouse.
Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex required for mitotic progression and cytokinesis. The BCR(KLHL42) E3 ubiquitin ligase complex mediates the ubiquitination and subsequent degradation of KATNA1. Involved in microtubule dynamics throughout mitosis. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ctb9; Cullin-3-binding protein 9; kelch domain containing 5; Kelch domain-containing protein 5; kelch-like family member 42; kelch-like family, member 42; Kelch-like protein 42
Gene Aliases: Ctb9; KIAA1340; KLHDC5; KLHL42
UniProt ID: (Human) Q9P2K6
Entrez Gene ID: (Human) 57542
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support