Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Defects in KLHL7 are the cause of retinitis pigmentosa type 42 (RP42). The specific function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BTB/POZ domain containing protein; kelch-like 6; kelch-like 7; kelch-like family member 7; Kelch-like protein 7; kelch/BTB
Gene Aliases: 2700038B03Rik; D5Ertd363e; KLHL6; KLHL7; SBBI26
UniProt ID: (Human) Q8IXQ5, (Mouse) Q8BUL5, (Rat) Q5XHZ6
Entrez Gene ID: (Human) 55975, (Mouse) 52323, (Rat) 362303
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support