Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • KLRG1 Antibodies

          Invitrogen

          KLRG1 Monoclonal Antibody (2F1), Brilliant Violet™ 480, eBioscience™

          View all (60) KLRG1 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite KLRG1 Monoclonal Antibody (2F1), Brilliant Violet™ 480, eBioscience™

          • Antibody Testing Data (1)
          KLRG1 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          KLRG1 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          KLRG1 Antibody (414-5893-82) in Flow

          C57BL/6 mouse splenocytes were stained with NK1.1 Monoclonal Antibody, PE (Product # 12-5941-82) and 0.25 µg of Syrian Hamster IgG Isotype Control, Brilliant Violet 480 (Product # 414-4914-81) (left) or 0.25 µg of KLRG1 Monoclonal Antibody, Brilliant Violet 480 (right). Viable cells in the lymphocyte gate were used for analysis, as determined by 7-AAD (Product # 00-6993-50). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          KLRG1 Antibody in Flow Cytometry (Flow)

          Product Details

          414-5893-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.5 µg/test
          -
          Product Specifications

          Species Reactivity

          Mouse

          Host/Isotype

          Syrian hamster / IgG

          Recommended Isotype Control

          Syrian Hamster IgG Isotype Control, Brilliant Violet™ 480, eBioscience™

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2F1

          Conjugate

          Brilliant Violet™ 480 Brilliant Violet™ 480 Brilliant Violet™ 480
          • Alexa Fluor 488 
          • Alexa Fluor 532 
          • Alexa Fluor 561
          • Alexa Fluor 647 
          • APC 
          • APC-eFluor 780
          • Biotin 
          • Brilliant UV 395
          • Brilliant UV 496
          • Brilliant UV 563
          • Brilliant UV 615
          • Brilliant Violet 421
          • eFluor 450
          • eFluor 506
          • FITC
          • Functional Grade
          • Pacific Orange
          • PE 
          • PE-Cyanine5
          • PE-Cyanine5.5
          • PE-Cyanine7
          • PE-eFluor 610
          • PerCP-eFluor 710
          • Super Bright 436
          • Super Bright 600
          • Super Bright 645
          • Super Bright 702
          • Super Bright 780
          • Request custom conjugation

          Excitation/Emission Max

          437/478 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2, with BSA

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_3093227

          Product Specific Information

          Description: This 2F1 monoclonal antibody reacts with the mouse Killer cell Lectin-like Receptor G1 (KLRG1), also known as Mast cell Function-associated Antigen (MAFA). KLRG1 is a homodimer of glycosylated 30-38 kDa subunits and contains a cytoplasmic motif similar to the immunoreceptor tyrosine-based inhibitory motif (ITIM). Rat MAFA was identified as an antigen specific to rat mast cells; however, the expression of mouse KLRG1/MAFA using 2F1 has not been detected on the surface of mouse mast cell lines, bone marrow-derived mast cells, or peritoneal mast cells. This antigen is expressed on approximately one-third of mouse NK cells and a subset of T cells. MHC class I molecules regulate KLRG1 via interactions with class I-specific inhibitory Ly49 molecules and SHP-1 signaling. Although KLRG1 and Ly49 are both lectin-like inhibitory receptors that are regulated by class I MHC expression, the effects of this on cell surface expression of these molecules are opposing, and the underlying regulatory mechanisms distinct.

          Applications Reported: This 2F1 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This 2F1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This may be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Brilliant Violet™ 480 (BV480) is a dye that emits at 479 nm and is intended for use on cytometers equipped with a violet (405 nm) laser. Please make sure that your instrument is capable of detecting this fluorochrome.

          When using two or more Super Bright, Brilliant Violet™, Brilliant Ultra Violet™, or other polymer dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Complete Staining Buffer (Product # SB-4401-42) or Brilliant Stain Buffer™ (Product # 00-4409-75) to minimize any non-specific polymer interactions. Please refer to the datasheet for Super Bright Staining Buffer or Brilliant Stain Buffer for more information.

          Our internal testing suggests that Brilliant Violet™ 480 (BV480) is not compatible with methanol-based fixation.

          Excitation: 440 nm; Emission: 479 nm; Laser: Violet Laser.

          BRILLIANT VIOLET™ is a trademark or registered trademark of Becton, Dickinson and Company or its affiliates, and is used under license. Powered by Sirigen™.

          Target Information

          KLRG1 (Killer cell lectin-like receptor G1, MAFA, 2F1-Ag) is a homodimeric member of the lectin-like type 2 transmembrane receptor family whose members contain characteristic immunoreceptor tyrosine-based inhibitory motifs (ITIMs) in their cytoplasmic domains. These ITIMs interact with the SH2 domains of protein phosphatases such as SHP-1. KLRG1 is an inhibitory receptor that is expressed on natural killer (NK) cells and certain T cells. NK cells are involved in the lysis of tumor cells and virus-infected cells, and mediate humoral and cell-mediated immune responses.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Killer cell lectin-like receptor subfamily G member 1; Mast cell function-associated antigen 2F1; natural killer cell receptor

          View more View less

          Gene Aliases: 2F1-Ag; Klrg1; MAFA; MAFA-L

          View more View less

          UniProt ID: (Mouse) O88713

          View more View less

          Entrez Gene ID: (Mouse) 50928

          View more View less

          Function(s)
          carbohydrate binding
          Process(es)
          immune system process cell surface receptor signaling pathway innate immune response
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0