Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EIVSIDTFN KSPPKRGLVV GITFIKDSGD KGSPFLNIYC DYEPGSEYNL DSIAQSCLNL ELQFTPFQLC HAEVQVGDQL ETVFLLSGND PAIHLYKENE GLHQFEEQPV ENLFPELTNL TSSVLWLDVH NFPGTSRRLS ALGCQSGYVR VAHVDQRSRE VLQMWSVLQD GPISRVIVFS LSAAKETKDR PLQDEYSVLV ASMLEPAVVY RDLLNRGLED QLLLPGSDQF DSVLCSLVTD VDLDGRPEVL VATYGQELLC YKYRGPESGL PEAQHGFHLL WQRSFSSPLL AMAHVDLTGD GLQELAVVSL KGVHILQHSL IQASELVLTR LRHQVEQRRR RLQGLEDGAG AGPAENAAS (79-436 aa encoded by BC009249)
Kaptin, also named as KPTN and Actin-associated protein 2E4, originally isolated from human blood platelets and likely to be involved in the actin rearrangements occurring during activation. It plays a unique role in the actin rearrangements that accompany platelet activation and stereocilia formation. Kaptin is necessary for normal neuromorphogenesis. It may play a role in producing the sensory apparatus in hair cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Actin-associated protein 2E4; actin-binding protein; kaptin; kaptin (actin binding protein); KICSTOR complex protein kaptin
Gene Aliases: 2310042D10Rik; 2E4; C030013F01Rik; KPTN; MRT41
UniProt ID: (Human) Q9Y664
Entrez Gene ID: (Human) 11133, (Mouse) 70394, (Rat) 308107
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support