Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MNSGHSFSQT PSASFHGAGG GWGRPRSFPR APTVHGGAGG ARISLSFTTR SCPPPGGSWG SGRSSPLLGG NGKATMQNLN DRLASYVEKV RALEEANMKL ESRILKWHQQ RDPGSKKDYS QYEENITHLQ EQIVDGKMTN AQIILLIDNA RMAVDDFNLK YENEHSFKKD LEIEVEGLRR TLDNLTIVTT DLEQEVEGMR KELILMKKHH EQEMEKHHVP SDFNVNVKVD TGPREDLIKV LEDMRQEYEL IIKKKHRDLD TWYKEQSAAM SQEAASPATV QSRQGDIHEL KRTFQALEID LQTQYSTKSA LENMLSETQS RYSCKLQDMQ EIISHYEEEL TQLRHELERQ NNEYQVLLGI KTHLEKEITT YRRLLEGESE GTREESKSSM KVFATPKIKA ITQETINGRL VLCQVNEIQK HA
The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. The type I cytokeratin genes are clustered in a region of chromosome 17q12-q21.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C HAIK1; CK-23; cytokeratin 23; Cytokeratin-23; DKFZP434G032; histone deacetylase inducible keratin 23; hyperacetylation-inducible type I keratin; K23; keratin 23 (histone deacetylase inducible); keratin 23, type I; Keratin, type I cytoskeletal 23; Keratin-23; MGC26158; type I intermediate filament cytokeratin
Gene Aliases: CK23; HAIK1; K23; KRT23
UniProt ID: (Human) Q9C075
Entrez Gene ID: (Human) 25984
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support