Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MASTSTTIRS HSSSRRGFSA NSARLPGVSR SGFSSISVSR SRGSGGLGGA CGGAGFGSRS LYGLGGSKRI SIGGGSCAIS GGYGSRAGGS YGFGGAGSGF GFGGGAGIGF GLGGGAGLAG GFGGPGFPVC PPGGIQEVTV NQSLLTPLNL QIDPAIQRVR AEEREQIKTL NNKFASFIDK VRFLEQQNKV LDTKWTLLQE QGTKTVRQNL EPLFEQYINN LRRQLDNIVG ERGRLDSELR NMQDLVEDLK NKYEDEINKR TAAENEFVTL KKDVDAAYMN KVELQAKADT LTDEINFLRA LYDAELSQMQ THISDTSVVL SMDNNRNLDL DSIIAEVKAQ YEEIAQRSRA EAESWYQTKY EELQITAGRH GDDLRNTKQE IAEINRMIQR LRSEIDHVKK QCANLQAAIA DAEQRGEMAL KDAKNKLEGL EDALQKAKQD LARLLKEYQE LMNVKLALDV EIATYRKLLE GEECRLNGEG VGQVNISVVQ STVSSGYGGA SGVGSGLGLG GGSSYSYGSG LGVGGGFSSS SGRATGGGLS SVGGGSSTIK YTTTSSSSRK SYKH
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. As many as six of this type II cytokeratin (KRT6) have been identified; the multiplicity of the genes is attributed to successive gene duplication events. The genes are expressed with family members KRT16 and/or KRT17 in the filiform papillae of the tongue, the stratified epithelial lining of oral mucosa and esophagus, the outer root sheath of hair follicles, and the glandular epithelia. Mutations in these genes have been associated with pachyonychia congenita. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-6B; cytokeratin 6B; Cytokeratin-6B; K6B; keratin 6B, type II; keratin, epidermal, type II, K6B; Keratin, type II cytoskeletal 6B; Keratin-6B; keratin-like 1 (a type II keratin sequence); Type-II keratin Kb10
Gene Aliases: CK-6B; CK6B; K6B; KRT6B; KRTL1; PC2; PC4
UniProt ID: (Human) P04259
Entrez Gene ID: (Human) 3854
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support