Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MRSSVSRQTY STKGGFSSNS ASGGSGSQAR TSFSSVTVSR SSGSGGGAHC GPGTGGFGSR SLYNLGGHKS ISVSVAGGAL LGRALGGFGF GSRAFMGQGA GRQTFGPACP PGGIQEVTVN QSLLTPLHVE IDPEIQRVRT QEREQIKTLN NKFASFIDKV RFLEQQNKVL ETKWALLQEQ GQNLGVTRNN LEPLFEAYLG SMRSTLDRLQ SERGRLDSEL RNVQDLVEDF KNKYEDEINK HTAAENEFVV LKKDVDAAYM GRMDLHGKVG TLTQEIDFLQ QLYEMELSQV QTHVSNTNVV LSMDNNRNLD LDSIIAEVKA QYELIAQRSR AEAEAWYQTK YEELQVTAGK HGDNLRDTKN EIAELTRTIQ RLQGEADAAK KQCQQLQTAI AEAEQRGELA LKDAQKKLGD LDVALHQAKE DLTRLLRDYQ ELMNVKLALD VEIATYRKLL ESEESRMSGE CPSAVSISVT GNSTTVCGGG AASFGGGISL GGSGGATKGG FSTNVGYSTV KGGPVSAGTS ILRKTTTVKT SSQRY
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes an epithelial keratin that is expressed in skeletal muscle, skin and scalp. The type II keratins are clustered in a region of chromosome 12q13.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-79; Cytokeratin-79; K79; keratin 6L; keratin 79, type II; Keratin, type II cytoskeletal 79; Keratin-6-like; Keratin-6L; Keratin-79; Type-II keratin Kb38
Gene Aliases: K6L; KB38; KRT6L; KRT79
UniProt ID: (Human) Q5XKE5
Entrez Gene ID: (Human) 338785
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support