Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MACRSCVVG FSSLSSCEVT PVGSPRPGTS GWDSCRAPGP GFSSRSLTGC WSAGTISKVT VNPGLLVPLD VKLDPAVQQL KNQEKEEMKA LNDKFASLIG KVQALEQRNQ LLETRWSFLQ GQDSAIFDLG HLYEEYQGRL QEELRKVSQE RGQLEANLLQ VLEKVEEFRI RYEDEISKRT DMEFTFVQLK KDLDAECLHR TELETKLKSL ESFVELMKTI YEQELKDLAA QVKDVSVTVG MDSRCHIDLS GIVEEVKAQY DAVAARSLEE AEAYSRSQLE EQAARSAEYG SSLQSSRSEI ADLNVRIQKL RSQILSVKSH CLKLEENIKT AEEQGELAFQ DAKTKLAQLE AALQQAKQDM ARQLRKYQEL MNVKLALDIE IATYRKLVEG EEGRMDSPSA TVVSAVQSRC KTAPSLPYPL CSL (1-422 aa encoded by BC065180)
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene's expression profile shows that it encodes a type II epithelial keratin, although structurally the encoded protein is more like a type II hair keratin. This protein is involved in cell differentiation, localizing near desmosomal plaques in earlier stages of differentiation but then dispersing throughout the cytoplasm in terminally differentiating cells. The type II keratins are clustered in a region of chromosome 12q13. Two transcript variants encoding two different fully functional isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CK-80; Cytokeratin-80; K80; keratin 80, type II; keratin b20; Keratin, type II cytoskeletal 80; Keratin-80; type II keratin; type II keratin Kb20; type II keratin-20; Type-II keratin Kb20
Gene Aliases: 1200016G03Rik; 2310041I20Rik; Kb2; KB20; KRT80
UniProt ID: (Human) Q6KB66, (Mouse) Q0VBK2, (Rat) Q6IMF1
Entrez Gene ID: (Human) 144501, (Mouse) 74127, (Rat) 315318
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support