Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MPYNFCLPSL SCRTSCSSRP CVPPSCHGYT LPGACNIPAN VSNCNWFCEG SFNGSEKETM QFLNDRLASY LEKVRQLERD NAELENLIRE RSQQQEPLLC PSYQSYFKTI EELQQKILCS KSENARLVVQ IDNAKLAADD FRTKYQTEQS LRQLVESDIN SLRRILDELT LCRSDLEAQM ESLKEELLSL KQNHEQEVNT LRCQLGDRLN VEVDAAPAVD LNQVLNETRN QYEALVETNR REVEQWFATQ TEELNKQVVS SSEQLQSYQA EIIELRRTVN ALEIELQAQH NLRYSLENTL TESEARYSSQ LSQVQSLITN VESQLAEIRS DLERQNQEYQ VLLDVRARLE CEINTYRSLL ESEDCKLPSN PCATTNACEK PIGSCVTNPC GPRSRCGPCN TFGY
The protein encoded by this gene is a member of the keratin gene family. It is one of the type I hair keratin genes which are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. There are two isoforms of this protein, encoded by two separate genes, KRTHA3A and KRTHA3B.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hair keratin; Hair keratin, type I Ha3-II; hard keratin, type I, 3II; K33B; keratin 33B, type I; keratin, hair, acidic, 3B; Keratin, type I cuticular Ha3-II; Keratin-33B; type I hair keratin 3B
Gene Aliases: Ha-3II; HA3II; HHA3-II; HKA3B; K33B; KRT33B; KRTHA3A; KRTHA3B
UniProt ID: (Human) Q14525
Entrez Gene ID: (Human) 3884
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support