Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide sequence: GEEAGMFSHV DMQPCGPLEG KANGGLVDGE VPELPPPLWA PPGKHLVTEV
Sequence homology: Cow: 79%; Dog: 86%; Guinea Pig: 93%; Human: 100%; Mouse: 93%; Rat: 100%
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: potassium channel, voltage gated shaker related subfamily A, member 7; Potassium voltage-gated channel subfamily A member 7; potassium voltage-gated channel, shaker-related subfamily, member 7; voltage-dependent potassium channel Kv1.7; voltage-gated potassium channel KCNA7; Voltage-gated potassium channel subunit Kv1.7
Gene Aliases: HAK6; KCNA7; KV1.7
UniProt ID: (Human) Q96RP8
Entrez Gene ID: (Human) 3743
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support