Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 7
Positive Control: Mouse brain tissue lysates, rat brain tissue lysates, rat cerebellum tissue lysates, EA.hy926, rat testis tissue, rat brain tissue, rat kidney tissue, rat cerebellum tissue, JAR.
Potassium channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. The KCNAB1 gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: K(+) channel subunit beta-1; K+ channel Beta1a chain; Kv-beta-1; potassium channel beta 3 chain; potassium channel beta3 subunit; potassium channel shaker chain beta 1a; potassium channel, voltage gated subfamily A regulatory beta subunit 1; potassium voltage gated channel shaker related subfamily beta member 1; potassium voltage gated channel, shaker related subfamily, beta member 1; potassium voltage-gated channel, shaker-related subfamily, beta member 1; voltage-gated potassium channel beta subunit; Voltage-gated potassium channel subunit beta-1
Gene Aliases: AKR6A3; Akr8a8; hKvb3; hKvBeta3; KCNA1B; KCNAB1; KV-BETA-1; Kvb1; Kvb1.3; Kvbeta1.1; KvBeta3; mKv(beta)1
UniProt ID: (Human) Q14722, (Mouse) P63143, (Rat) P63144
Entrez Gene ID: (Human) 7881, (Mouse) 16497, (Rat) 29737
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support