Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Exalpha
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive control: Extraembryonic membranes from chick embryos.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 58 kDa cytokeratin; Cell proliferation-inducing gene 46 protein; CK-10; CK-13; CK-18; CK-4; CK-5; CK-8; cytokeratin 10; cytokeratin 13; cytokeratin 18; cytokeratin 4; Cytokeratin-10; Cytokeratin-13; Cytokeratin-18; Cytokeratin-4; Cytokeratin-5; Cytokeratin-8; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K10; K13; K18; K4; K5; K8; keratin 10, type I; keratin 13, type I; keratin 18, type I; keratin 4, type II; keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 8, type II; Keratin, type I cytoskeletal 10; Keratin, type I cytoskeletal 13; Keratin, type I cytoskeletal 18; Keratin, type II cytoskeletal 4; Keratin, type II cytoskeletal 5; Keratin, type II cytoskeletal 8; Keratin-10; Keratin-13; Keratin-18; Keratin-4; Keratin-5; Keratin-8; Type-II keratin Kb4; Type-II keratin Kb5; Type-II keratin Kb8
Gene Aliases: BCIE; BIE; CARD2; CK-18; CK-4; CK-8; CK10; CK13; CK4; CK5; CK8; CYK18; CYK4; CYK8; DDD; DDD1; EBS2; EHK; K10; K13; K18; K2C8; K4; K5; K8; KO; KPP; KRT10; KRT13; KRT18; KRT4; KRT5; KRT5A; KRT8; PIG46; WSN1; WSN2
UniProt ID: (Human) P13645, (Human) P13646, (Human) P05783, (Human) P19013, (Human) P13647, (Human) P05787
Entrez Gene ID: (Human) 3858, (Human) 3860, (Human) 3875, (Human) 3851, (Human) 3852, (Human) 3856
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support