Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LENAGCNKI GNFSTDIKDS KAYYHLLEQV APKGDEEGVP AVVIDMSGLR EKDDIQRAEC MLQQAERLGC RQFVTATDVV RGNPKLNLAF IANLFNRYPA LHKPENQDID WGALEGETRE ERTFRNWMNS LGVNPRVNHL YSDLSDALVI FQLYEKIKVP VDWNRVNKPP YPKLGGNMKK LENCNYAVEL GKNQAKFSLV GIGGQDLNEG NRTLTLALIW QLMRRYTLNI LEEIGGGQKV NDDIIVNWVN ETLREAEKSS SISSFKDPKI STSLPVLDLI DAIQPGSINY DLLKTENLND DEKLNNAKYA ISMARKIGAR VYALPEDLVE VNPKMVMTVF ACLMGKGMKR V (278-627 aa encoded by BC010271)
L-plastin is a leukocyte specific actin bundling protein from fimbrin family of proteins. It has two actin-binding domains and two EF hand type calcium-binding domains. It gets phosphorylated on the serine residue in its headpiece region in response to a variety of inflammatory stimuli. It is hypothesized that L-plastin may play a role in regulating integrin mediated adhesion in leukocytes. L-plastin is expressed in majority of human cancer cell lines derived from solid tumors. Its expression is positively upregulated by testosterones in androgen receptor positive prostrate and breast cancer cells through three androgen receptor binding elements located in the upstream region of the gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 65 kDa macrophage protein; bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)); DKFZp781A23186; epididymis secretory protein Li 37; FLJ25423; FLJ26114; FLJ39956; L-fimbrin; L-plastin; L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (65 kDa macrophage protein) (PP65); L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P); LC64P; LCP-1; Lymphocyte cytosolic protein 1; lymphocyte cytosolic protein 1 (L-plastin); Lymphocyte cytosolic protein-1 (plasmin); plastin 2; plastin 2, L; Plastin-2; pp65
Gene Aliases: AW536232; CP64; D14Ertd310e; HEL-S-37; L-PLASTIN; LC64P; LCP-1; LCP1; LPL; PLS2; pp65
UniProt ID: (Human) P13796, (Mouse) Q61233
Entrez Gene ID: (Human) 3936, (Mouse) 18826, (Rat) 306071
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support