Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Histone chaperone that specifically mediates the genome-wide removal of histone H2A.Z/H2AZ1 from the nucleosome: removes H2A.Z/H2AZ1 from its normal sites of deposition, especially from enhancer and insulator regions. Not involved in deposition of H2A.Z/H2AZ1 in the nucleosome. May stabilize the evicted H2A.Z/H2AZ1-H2B dimer, thus shifting the equilibrium towards dissociation and the off-chromatin state (PubMed:24463511). Inhibits activity of protein phosphatase 2A (PP2A). Does not inhibit protein phosphatase 1. May play a role in cerebellar development and synaptogenesis. [Uniprot]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: acidic (leucine-rich) nuclear phosphoprotein 32 family, member E; Acidic leucine-rich nuclear phosphoprotein 32 family member E; Cerebellar postnatal development protein 1; LANP-L; LANP-like protein; leucine-rich acidic nuclear protein like
Gene Aliases: 2810018A15Rik; AI047746; AI326868; ANP32E; CPD1; LANP-L; LANPL; mLANP-L
UniProt ID: (Human) Q9BTT0, (Mouse) P97822, (Rat) Q5XIE0
Entrez Gene ID: (Human) 81611, (Mouse) 66471, (Rat) 361999
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support