Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody can recognize all the 3 isoforms of LAPTM4B.
Immunogen sequence: TDPAEARSS RGIEEAGPRA HGRAGREPER RRSRQQRRGG LQARRSTLLK TCARARATAP GAMKMVAPWT RFYSNSCCLC CHVRTGTILL GVWYLIINAV VLLILLSALA DPDQYNFSSS ELGGDFEFMD DANMCIAIAI SLLMILICAM ATYGAYKQRA AWIIPFFCYQ IFDFALNMLV AITVLIYPNS IQEYIRQLPP NFPYRDDVMS VNPTCLVLII LLFISIILTF KGYLISCVWN CYRYINGRNS SDVLVYVTSN DTTVLLPPYD DATVNGAAKE PPPPYVSA (84-370 aa encoded by BC014129 )
LAPTM4B is a multi-pass membrane protein. It belongs to the LAPTM4/LAPTM5 transporter family. LAPTM4b has active role in disease progression of malignant cells and is involved in cell proliferation and multidrug resistance. The genetic polymorphism of LAPTM4B is a potential risk factor for the development of colon cancer and gastric cancer. The research results also indicated that LAPTM4B may be a clinically useful prognostic indicator for ovarian carcinoma and may play a role in human hepatocellular carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: lysosomal associated protein transmembrane 4 beta; lysosomal-associated protein transmembrane 4B; lysosomal-associated transmembrane protein 4; Lysosomal-associated transmembrane protein 4B; Lysosome-associated transmembrane protein 4-beta
Gene Aliases: C330023P13Rik; LAPTM4B; LAPTM4beta; LC27; PSEC0001
UniProt ID: (Human) Q86VI4, (Rat) Q5U1W4, (Mouse) Q91XQ6
Entrez Gene ID: (Human) 55353, (Rat) 315047, (Mouse) 114128
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support