Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • LBP Antibodies

          Invitrogen

          LBP Polyclonal Antibody

          View all (12) LBP antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite LBP Polyclonal Antibody

          • Antibody Testing Data (1)
          LBP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          LBP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LBP Antibody (PA5-79584) in WB

          Western blot analysis of LBP in rat thymus extract (lane 1), mouse thymus extract (lane 2), HeLa whole cell lysate (lane 3) and K562 whole cell lysate (lane 4). Sample was incubated with LBP polyclonal antibody (Product # PA5-79584) at a dilution of 0.5 µg/mL. Signal development was performed using a chemiluminescence (ECL) kit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LBP Antibody in Western Blot (WB)

          Product Details

          PA5-79584

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived human LBP recombinant protein (Position: L177-E446).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 5mg BSA

          Contains

          0.05mg sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2746699

          Product Specific Information

          Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

          Positive Control - WB: rat thymus tissue, mouse thymus tissue, HELA whole cell, K562 whole cell.

          Target Information

          The LBP protein is involved in the acute-phase immunologic response to gram-negative bacterial infections. Gram-negative bacteria contain a glycolipid, lipopolysaccharide (LPS), on their outer cell wall. Together with bactericidal permeability-increasing protein (BPI), the encoded protein binds LPS and interacts with the CD14 receptor, probably playing a role in regulating LPS-dependent monocyte responses. This protein is part of a family of structurally and functionally related proteins, including BPI, plasma cholesteryl ester transfer protein (CETP), and phospholipid transfer protein (PLTP). Finally, this gene is found on chromosome 20, immediately downstream of the BPI gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BPI fold containing family D, member 2; LBP; Lipopolysaccharide-binding protein; LPS-binding protein

          View more View less

          Gene Aliases: BPIFD2; LBP; Ly88

          View more View less

          UniProt ID: (Human) P18428, (Mouse) Q61805

          View more View less

          Entrez Gene ID: (Human) 3929, (Mouse) 16803, (Rat) 29469

          View more View less

          Function(s)
          lipopolysaccharide binding receptor binding protein binding lipoteichoic acid binding lipid binding
          Process(es)
          liver development leukocyte chemotaxis involved in inflammatory response macrophage activation involved in immune response acute-phase response cellular defense response opsonization lipopolysaccharide transport lipopolysaccharide-mediated signaling pathway detection of molecule of bacterial origin response to lipopolysaccharide negative regulation of tumor necrosis factor production positive regulation of chemokine production positive regulation of interleukin-6 production positive regulation of interleukin-8 production positive regulation of tumor necrosis factor production toll-like receptor 4 signaling pathway positive regulation of toll-like receptor 4 signaling pathway positive regulation of tumor necrosis factor biosynthetic process positive regulation of macrophage activation negative regulation of growth of symbiont in host innate immune response defense response to Gram-negative bacterium defense response to Gram-positive bacterium positive regulation of phagocytosis, engulfment positive regulation of respiratory burst involved in inflammatory response cellular response to lipopolysaccharide cellular response to lipoteichoic acid positive regulation of neutrophil chemotaxis regulation of membrane permeability immune system process transport lipid transport defense response to bacterium
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0