Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May play a role in male fertility. May act as a retinoid carrier protein within the epididymis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Epididymal-specific lipocalin-8; lipocalin 5
Gene Aliases: EP17; LCN5; LCN8
UniProt ID: (Human) Q6JVE9
Entrez Gene ID: (Human) 138307
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support