Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSSTPHDPFY SSPFGPFYRR HTPYMVQPEY RIYEMNKRLQ SRTEDSDNLW WDAFATEFFE DDATLTLSFC LEDGPKRYTI GRTLIPRYFS TVFEGGVTDL YYILKHSKES YHNSSITVDC DQCTMVTQHG KPMFTKVCTE GRLILEFTFD DLMRIKTWHF TIRQYRELVP RSILAMHAQD PQVLDQLSKN ITRMGLTNFT LNYLRLCVIL EPMQELMSRH KTYNLSPRDC LKTCLFQKWQ RMVAPPAEPT RQPTTKRRKR KNSTSSTSNS SAGNNANSTG SKKKTTAANL SLSSQVPDVM VVGEPTLMGG EFGDEDERLI TRLENTQYDA ANGMDDEEDF NNSPALGNNS PWNSKPPATQ ETKSENPPPQ ASQ
Genes encoding LIM domain-binding factors were initially isolated in a screen for proteins that physically interact with the LIM domains of nuclear proteins. These proteins, such as the one encoded by the LDB2 gene, are capable of binding to a variety of transcription factors and are likely to function at enhancers to bring together diverse transcription factors and form higher order activation complexes or to block formation of such complexes. The family of genes encoding LIM domain-binding factors includes 2 members isolated from the mouse, Clim1 and Clim2/Lbd1/Nli; and their homologs cloned from the frog, chicken, and fly. The fact that LIM domain-binding factors are likely to be involved in the coordination of the transcriptional activity of many diverse factors might implicate them in human phenotypes characterized by multiple affected sites.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Carboxyl-terminal LIM domain-binding protein 1; CLIM-1; LDB-2; LIM binding domain 2; LIM domain-binding factor CLIM1; LIM domain-binding factor-2; LIM domain-binding protein 2
Gene Aliases: CLIM1; LDB-2; LDB1; LDB2
UniProt ID: (Human) O43679
Entrez Gene ID: (Human) 9079
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support