Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VNDGDMRLA DGGATNQGRV EIFYRGQWGT VCDNLWDLTD ASVVCRALGF ENATQALGRA AFGQGSGPIM LDEVQCTGTE ASLADCKSLG WLKSNCRHER DAGVVCTNET RSTHTLDLSR ELSEALGQIF DSQRGCDLSI SVNVQGEDAL GFCGHTVILT ANLEAQALWK EPGSNVTMSV DAECVPMVRD LLRYFYSRRI DITLSSVKCF HKLASAYGAR QLQGYCASLF AILLPQDPSF QMPLDLYAYA VATGDALLEK LCLQFLAWNF EALTQAEAWP SVPTDLLQLL LPRSDLAVPS ELALLKAVDT WSWGERASHE EVEGLVEKIR FPMMLPEELF ELQFNLSLYW SHEALFQKKT LQALEFHTVP FQLLARYKGL NLTEDTYKPR IYTSPTWSAF VTDSSWSARK SQLVYQSRRG PLVKYSSDYF QAPSDYRYYP YQSFQTPQHP SFLFQDKRVS WSLVYLPTIQ SCWNYGFSCS SDELPVLGLT KSGGSDRTIA YENKALMLCE GLFVADVTDF EGWKAAIPSA LDTNSSKSTS SFPCPAGHFN GFRTVIRPFY LTNSSGVD (19-585 aa encoded by BC002403 )
The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS3BP has been found elevated in the serum of patients with cancer and in those infected by the human immunodeficiency virus (HIV). It appears to be implicated in immune response associated with natural killer (NK) and lymphokine-activated killer (LAK) cell cytotoxicity. Using fluorescence in situ hybridization the full length 90K cDNA has been localized to chromosome 17q25. The native protein binds specifically to a human macrophage-associated lectin known as Mac-2 and also binds galectin 1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Basement membrane autoantigen p105; CyCAP; Cyp-C-associated protein; Galectin-3-binding protein; L3 antigen; Lectin; lectin galactoside-binding soluble 3 binding protein; Lectin galactoside-binding soluble 3-binding protein; lectin, galactoside binding soluble 3 binding protein; lectin, galactoside-binding, soluble, 3 binding protein; mac-2 BP; Mac-2-binding protein; MAC2BP; mama; peptidylprolyl isomerase C-associated protein; Protein MAMA; serum protein 90K; transport and golgi organization 10 homolog B; Tumor-associated antigen 90K
Gene Aliases: 90K; BTBD17B; CyCAP; gp90; LGALS3BP; M2BP; MAC-2-BP; MAC-2BP; Mama; Ppicap; TANGO10B
UniProt ID: (Human) Q08380, (Mouse) Q07797
Entrez Gene ID: (Human) 3959, (Mouse) 19039
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support