Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MAATLGPLG SWQQWRRCLS ARDGSRMLLL LLLLGSGQGP QQVGAGQTFE YLKREHSLSK PYQGVGTGSS SLWNLMGNAM VMTQYIRLTP DMQSKQGALW NRVPCFLRDW ELQVHFKIHG QGKKNLHGDG LAIWYTKDRM QPGPVFGNMD KFVGLGVFVD TYPNEEKQQE RVFPYISAMV NNGSLSYDHE RDGRPTELGG CTAIVRNLHY DTFLVIRYVK RHLTIMMDID GKHEWRDCIE VPGVRLPRGY YFGTSSITGD LSDNHDVISL KLFELTVERT PEEEKLHRDV FLPSVDNMKL PEMTAPLPPL SGLALF (1-315 aa encoded by B C067265)
May be involved in the regulation of export from the endoplasmic reticulum of a subset of glycoproteins. May function as a regulator of ERGIC-53.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lectin mannose-binding 2-like; LMAN2-like protein; VIP36-like protein
Gene Aliases: A630028F14Rik; LMAN2L; MRT52; PSEC0028; UNQ368/PRO704; VIP36-like; VIPL
UniProt ID: (Human) Q9H0V9, (Mouse) P59481
Entrez Gene ID: (Human) 81562, (Mouse) 214895
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support