Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MVLLERLRHP NVLQLYGYCY QDSEDIPDTL TTITELGAPV EMIQLLQTSW EDRFRICLSL GRLLHHLAHS PLGSVTLLDF RPRQFVLVDG ELKVTDLDDA RVEETPCAGS TDCILEFPAR NFTLPCSAQG WCEGMNEKRN LYNAYRFFFT YLLPHSAPPS LRPLLDSIVN ATGELAWGVD ETLAQLEKVL HLYRSGQYLQ NSTASSSTEY QCIPDSTIPQ EDYRCWPSYH HGSCLLSVFN LAEAVDVCES HAQCRAFVVT NQTTWTGRQL VFFKTGWSQV VPDPNKTTYV KASG
VLK was identified as a novel protein kinase that was induced after the differentiation of cultured embryonic stem cells into mesendoderm. It has no homologs in invertebrates, but is highly conserved in vertebrate species although it does not belong to any known protein kinase groups. VLK is initially expressed in E-cadherin-positive anterior visceral endoderm and mesendoderm, but its expression is later confined to E-cadherin-negative mesenchyme. It is enriched in the Golgi apparatus and is thought to regulate the rate of protein export from the Golgi. Targeted disruption of VLK in mice leads to a defect in lung development and neonatal lethality. It has been suggested that mutations in VLK may be associated with the allergic condition atopy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Extracellular tyrosine-protein kinase PKDCC; protein kinase domain containing, cytoplasmic homolog; Protein kinase domain-containing protein, cytoplasmic; Protein kinase-like protein SgK493; Sugen kinase 493; Vertebrate lonesome kinase
Gene Aliases: PKDCC; SGK493; VLK
UniProt ID: (Human) Q504Y2
Entrez Gene ID: (Human) 91461
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support