Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MSYQKKQPTP QPPVDCVKTS GGGGGGGGGG GGGCGFFGGG GSGGGSSGSG CGYSGGGGYS GGGCGGGSSG GGGGGGIGGC GGGSGGSVKY SGGGGSSGGG SGCFSSGGGG SGCFSSGGGG SSGGGSGCFS SGGGGSSGGG SGCFSSGGGG FSGQAVQCQS YGGVSSGGSS GGGSGCFSSG GGGGSVCGYS GGGSGGGSGC GGGSSGGSGS GYVSSQQVTQ TSCAPQPSYG GGSSGGGGSG GSGCFSSGGG GGSSGCGGGS SGIGSGCIIS GGGSVCGGGS SGGGGGGSSV GGSGSGKGVP ICHQTQQKQA PTWPSK
This gene encodes loricrin, a major protein component of the cornified cell envelope found in terminally differentiated epidermal cells. Mutations in this gene are associated with Vohwinkel's syndrome and progressive symmetric erythrokeratoderma, both inherited skin diseases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Loricrin; MGC111513
Gene Aliases: LOR; LORICRIN; LRN
UniProt ID: (Human) P23490
Entrez Gene ID: (Human) 4014
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support