Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17111 targets LPA in indirect ELISA, ICC, IHC, IF and WB applications and shows reactivity with Human samples.
The MA5-17111 immunogen is purified recombinant fragment of human LPA (amino acids: 4330-4521) expressed in E. Coli.
MA5-17111 detects LPA which has a predicted molecular weight of approximately 501kDa.
The protein encoded by this gene is a serine proteinase that inhibits the activity of tissue-type plasminogen activator I. The encoded protein constitutes a substantial portion of lipoprotein(a) and is proteolytically cleaved, resulting in fragments that attach to atherosclerotic lesions and promote thrombogenesis. Elevated plasma levels of this protein are linked to atherosclerosis. Depending on the individual, the encoded protein contains 2-43 copies of kringle-type domains. The allele represented here contains 15 copies of the kringle-type repeats and corresponds to that found in the reference genome sequence.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: antiangiogenic AK38 protein; Apo(a); Apolipoprotein(a); lipoprotein, Lp(a); lp(a)
Gene Aliases: AK38; APOA; LP; LPA
UniProt ID: (Human) P08519
Entrez Gene ID: (Human) 4018
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support