Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Alomone Labs, LTD
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C, protected from the light, for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 X g 5 min).
EDG7 is a member of the G protein-coupled receptor family, as well as the EDG family of proteins. This protein functions as a cellular receptor for lysophosphatidic acid and mediates lysophosphatidic acid-evoked calcium mobilization. This receptor couples predominantly to G(q/11) alpha proteins.EDG7 has been reported in human prostate, as well as rodent brain, kidney, lung, placenta, prostate, and testis. ESTs have been isolated from a normal prostate library, as well as cancer libraries of the head/neck, ovary, and thyroid.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: calcium-mobilizing lysophosphatidic acid receptor LP-A3; endothelial cell differentiation gene 7; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor 7; endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor, 7; LPA receptor 3; LPA receptor EDG7; LPA-3; LPA3 Receptor; Lysophosphatidic acid receptor 3; Lysophosphatidic acid receptor Edg-7; putative G protein-coupled receptor snGPCR32; snGPCR32
Gene Aliases: Edg-7; EDG7; GPCR; HOFNH30; LP-A3; LPA3; LPAR3; RP4-678I3
UniProt ID: (Human) Q9UBY5, (Mouse) Q9EQ31, (Rat) Q8K5E0
Entrez Gene ID: (Human) 23566, (Mouse) 65086, (Rat) 66025
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support