Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
LRCH3 (Leucine Rich Repeats And Calponin Homology Domain Containing 3) is a Protein Coding gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DISP complex protein LRCH3; Leucine-rich repeat and calponin homology domain-containing protein 3; leucine-rich repeats and calponin homology (CH) domain containing 3; MGC4126
Gene Aliases: 2210409B11Rik; AW215594; Gm1742; LRCH3
UniProt ID: (Human) Q96II8, (Mouse) Q8BVU0
Entrez Gene ID: (Human) 84859, (Rat) 303883, (Mouse) 70144
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support