Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • LRP5 Antibodies

          AbboMax

          LRP5 Polyclonal Antibody

          View all (12) LRP5 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite LRP5 Polyclonal Antibody

          • Antibody Testing Data (1)
          LRP5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          LRP5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LRP5 Antibody (601-020) in WB

          Western Blot: The cell lysate derived from 3T3 was immunoprobed by Rabbit anti-LRP5 (Cat#601-020) at 1:500. An immunoreactive band is observed around ~175 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LRP5 Antibody in Western Blot (WB)

          Product Details

          601-020

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-1 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          2-10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide corresponding to the epitope KATLY of human LRP5 protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with proprietary stabilizer

          Contains

          0.01% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Product Specific Information

          Positive control: LRP5 transfected HEK293

          Cellular location: Membrane, Endoplasmic reticulum.

          Target Information

          LDL related proteins, LRP 5 and LRP 6, are members of the LDL receptor superfamily, which were found to be involved in the canonical Wnt signaling pathway. Mammalian LRP 6 was shown to bind to Wnt 1 and enhance Wnt-induced developmental processes in Xenopus embryos. Mice lacking LRP 6 exhibited developmental defects that are similar to those caused by deficiencies in various Wnt proteins. Recent work also shows that Wnt proteins induce the binding of LRP 5 to Axin, leading Axin degradation and beta catenin stabilization.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HGNC:6697; low density lipoprotein receptor-related protein 5; low density lipoprotein receptor-related protein 7; Low-density lipoprotein receptor-related protein 5; Low-density lipoprotein receptor-related protein 7; LRP-5; LRP-7

          View more View less

          Gene Aliases: BMND1; EVR1; EVR4; HBM; LR3; LRP-5; LRP5; LRP7; mKIAA4142; OPPG; OPS; OPTA1; VBCH2

          View more View less

          UniProt ID: (Human) O75197, (Mouse) Q91VN0

          View more View less

          Entrez Gene ID: (Human) 4041, (Rat) 293649, (Mouse) 16973

          View more View less

          Function(s)
          protein binding Wnt-protein binding toxin transporter activity Wnt-activated receptor activity coreceptor activity involved in Wnt signaling pathway coreceptor activity involved in canonical Wnt signaling pathway molecular_function coreceptor activity
          Process(es)
          gastrulation with mouth forming second positive regulation of mesenchymal cell proliferation osteoblast development glucose catabolic process endocytosis cholesterol metabolic process regulation of blood pressure positive regulation of cell proliferation anterior/posterior pattern specification Wnt signaling pathway positive regulation of osteoblast proliferation somatic stem cell population maintenance extracellular matrix-cell signaling cell migration involved in gastrulation cholesterol homeostasis embryonic digit morphogenesis regulation of apoptotic process Wnt signaling pathway involved in dorsal/ventral axis specification positive regulation of fat cell differentiation negative regulation of osteoblast differentiation positive regulation of mitotic nuclear division positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter bone remodeling regulation of bone remodeling bone marrow development positive regulation of sequence-specific DNA binding transcription factor activity anatomical structure regression retina morphogenesis in camera-type eye canonical Wnt signaling pathway bone morphogenesis branching involved in mammary gland duct morphogenesis adipose tissue development cell-cell signaling involved in mammary gland development regulation of canonical Wnt signaling pathway regulation of insulin secretion involved in cellular response to glucose stimulus retinal blood vessel morphogenesis negative regulation of protein serine/threonine kinase activity apoptotic process involved in patterning of blood vessels beta-catenin destruction complex disassembly vasculature development regulation of ossification embryonic limb morphogenesis limb morphogenesis response to peptide hormone regulation of fat cell differentiation embryonic camera-type eye morphogenesis embryonic retina morphogenesis in camera-type eye bone development mammary gland duct morphogenesis retina vasculature morphogenesis in camera-type eye multicellular organism development
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0