Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Auxiliary protein of the large-conductance, voltage and calcium-activated potassium channel (BK alpha). Modulates gating properties by producing a marked shift in the BK channel's voltage dependence of activation in the hyperpolarizing direction, and in the absence of calcium.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BK channel auxiliary gamma subunit LRRC38; BK channel auxilliary gamma subunit LRRC38; Leucine-rich repeat-containing protein 38
Gene Aliases: A230053A07Rik; LRRC38
UniProt ID: (Human) Q5VT99, (Mouse) A2VDH3
Entrez Gene ID: (Human) 126755, (Mouse) 242735
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support