Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
A suggested positive control is rat colon lysate.
PA5-21115 can be used with blocking peptide PEP-1229.
The leucine-rich repeat FLI-I-interacting protein 2 (LRRFIP2), like the related protein LRRFIP1, was identified in a yeast two-hybrid system through binding to the LRR domain of human FLI. It can activate the Wnt signaling pathway in cultured cells and is thought to be a component of the Wnt signaling pathway that modulates Wnt signaling through interactions with Disheveled to increase the cellular levels and transcription activity of beta-catenin. LRRFIP2 has recently been characterized as a positive regulator of the TLR4 signaling pathway for activating NF-kappa-B during the early host response to LPS stimulation through binding to the TLR adaptor protein MyD88, and that this interaction with MyD88 is governed by phosphorylation of specific residues in LRRFIP2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: leucine rich repeat (in FLII) interacting protein 2; Leucine-rich repeat flightless-interacting protein 2; LRR FLII-interacting protein 2
Gene Aliases: 5133400F20Rik; AI850587; HUFI-2; LRRFIP2
UniProt ID: (Human) Q9Y608, (Rat) Q4V7E8, (Mouse) Q91WK0
Entrez Gene ID: (Human) 9209, (Rat) 301035, (Mouse) 71268
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support