Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene is a neuronal surface glycoprotein found in cortical and subcortical regions of the limbic system. During development of the limbic system, this encoded protein is found on the surface of axonal membranes and growth cones, where it acts as a selective homophilic adhesion molecule, and guides the development of specific patterns of neuronal connections.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IgLON family member 3; Limbic system-associated membrane protein; LSAMP
Gene Aliases: 5430428I19; AW046674; B130007O04Rik; D930023J12Rik; IGLON3; Lam; LAMP; LSAMP
UniProt ID: (Human) Q8IV49, (Rat) Q62813
Entrez Gene ID: (Human) 4045, (Mouse) 268890, (Rat) 29561
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support