Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: FIEKKKAVSF HLVHRSQRDP LAADESAPQR VLLPTQKIDN EERRAEQRKY GVFFDDDYDY LQHLKEPSGP SELIPSSTFS AHNRREEKEE TLVIPS
Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 82%.
LTV1 gene ontology annotations related to this gene include ribosomal small subunit biogenesis; ribosomal small subunit export from nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LTV1 homolog; Protein LTV1 homolog
Gene Aliases: C6orf93; dJ468K18.4; LTV1
UniProt ID: (Human) Q96GA3
Entrez Gene ID: (Human) 84946
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support