Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This target displays homology in the following species: Dog: 93%; Horse: 79%; Human: 100%; Pig: 79%
LY6G5B belongs to a cluster of leukocyte antigen-6 (LY6) genes located in the major histocompatibility complex (MHC) class III region on chromosome 6. Members of the LY6 superfamily typically contain 70 to 80 amino acids, including 8 to 10 cysteines. Most LY6 proteins are attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor that is directly involved in signal transduction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lymphocyte antigen 6 complex locus protein G5b; lymphocyte antigen-6 G5B splicing isoform 288; lymphocyte antigen-6 G5B splicing isoform 452; lymphocyte antigen-6 G5B splicing isoform 690; lymphocyte antigen-6 G5B splicing isoform 837
Gene Aliases: C6orf19; G5B; LY6G5B
UniProt ID: (Human) Q5SQ62
Entrez Gene ID: (Human) 58496
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support