Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of total Lymphocyte Antigen 6 Complex.
Required for sperm migration into the oviduct and male fertility by controlling binding of sperm to zona pellucida. May play a role in cell growth (PubMed:18089789). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer/testis antigen 97; FLJ35226; Ly-6K; Lymphocyte antigen 6K; up-regulated in lung cancer 10
Gene Aliases: 2410015A16Rik; 3110035B01Rik; CO16; CT97; HSJ001348; ly-6K; LY6K; mLy-6K; URLC10
UniProt ID: (Human) Q17RY6, (Mouse) Q9CWP4
Entrez Gene ID: (Human) 54742, (Mouse) 76486
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support