Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: THDALPNLSQ APDPPTLSGA ECYACIGVHQ DDCAIGRSRR VQCHQDQTAC FQGSGRMTVG NFSVPVYIRT CHRPSCTTEG TTSPWTAIDL QGSCCEGYLC NRKSMTQPFT
The specific function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ly6/PLAUR domain-containing protein 5; metastasis-associated protein
Gene Aliases: LYPD5; PRO4356; UNQ1908/PRO4356
UniProt ID: (Human) Q6UWN5
Entrez Gene ID: (Human) 284348
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support