Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: TEGLVEATMP PGVQLHCLYG TGVPTPDSFY YESFPDRDPK ICFGDGDGTV NLKSALQCQA WQSRQEHQVL LQELPGSEHI EMLANATTLA YLKRVLLGP
Lysophospholipases are enzymes that act on biological membranes to regulate the multifunctional lysophospholipids. The protein encoded by this gene hydrolyzes lysophosphatidylcholine to glycerophosphorylcholine and a free fatty acid. This enzyme is present in the plasma and thought to be associated with high-density lipoprotein. A later paper contradicts the function of this gene. It demonstrates that this gene encodes a lysosomal enzyme instead of a lysophospholipase and has both calcium-independent phospholipase A2 and transacylase activities.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 1-O-acylceramide synthase; 1-O-acylceramide synthase {ECO:0000303|PubMed:15294901}; ACS; ACS {ECO:0000303|PubMed:15294901}; group XV phospholipase A2; group XV phospholipase A2 {ECO:0000312|EMBL:AAH98894.1}; LCAT-like lysophospholipase; LCAT-like lysophospholipase {ECO:0000250|UniProtKB:Q8VEB4}; LLPL; LLPL {ECO:0000250|UniProtKB:Q8VEB4}; LPLA2; LPLA2 {ECO:0000303|PubMed:15294901}; Lysophospholipase 3; lysophospholipase 3 (lysosomal phospholipase A2); lysophospholipase 3 {ECO:0000250|UniProtKB:Q8VEB4}; Lysosomal phospholipase A and acyltransferase; Lysosomal phospholipase A2; lysosomal phospholipase A2 {ECO:0000303|PubMed:15294901, ECO:0000312|EMBL:AAS66767.1}; Phospholipase A2 group XV; phospholipase A2, group XV; PLA2G15; pla2g15 {ECO:0000312|Ensembl:ENSRNOP00000026996, ECO:0000312|RGD:1302982}
Gene Aliases: 1-O-acylceramidesynthase; ACS; C87498; GXVPLA2; LLPL; LPLA2; LYPLA3; PLA2G15; UNQ341/PRO540
UniProt ID: (Human) Q8NCC3, (Mouse) Q8VEB4, (Rat) Q675A5
Entrez Gene ID: (Human) 23659, (Mouse) 192654, (Rat) 361401
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support