Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MSTDSNSLAR EFLTDVNRLC NAVVQRVEAR EEEEEETHMA TLGQYLVHGR GFLLLTKLNS IIDQALTCRE ELLTLLLSLL PLVWKIPVQE EKATDFNLPL
May be required for sorting endosomal resident proteins into late multivesicular endosomes by a mechanism involving microtubules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Beige homolog; Chediak-Higashi syndrome 1; Lysosomal-trafficking regulator
Gene Aliases: CHS; CHS1; LYST
UniProt ID: (Human) Q99698
Entrez Gene ID: (Human) 1130
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support