Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
LZIC belongs to the CTNNBIP1 family. It is ubiquitously expressed with highest levels in kidney. LZIC is up-regulated in several cases of gastric cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: leucine zipper and CTNNBIP1 domain containing; Leucine zipper and CTNNBIP1 domain-containing protein; Leucine zipper and ICAT homologous domain-containing protein; leucine zipper domain and ICAT homologous domain containing; Protein LZIC
Gene Aliases: 1810030J04Rik; AW047580; LZIC
UniProt ID: (Human) Q8WZA0, (Rat) Q5PQN7, (Mouse) Q8K3C3
Entrez Gene ID: (Dog) 100855658, (Human) 84328, (Rat) 366507, (Mouse) 69151
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support