Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • LZTFL1 Antibodies

          Proteintech

          LZTFL1 Monoclonal Antibody (2C1F6)

          View all (17) LZTFL1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite LZTFL1 Monoclonal Antibody (2C1F6)

          • Antibody Testing Data (3)
          LZTFL1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          LZTFL1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          LZTFL1 Antibody (68281-1-IG) in ICC/IF

          Immunofluorescent analysis of (4% PFA) fixed Jurkat cells using LZTFL1 antibody (68281-1-Ig, Clone: 2C1F6 ) at dilution of 1:800 and CoraLite®488-Conjugated AffiniPure Goat Anti-Mouse IgG(H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          LZTFL1 Antibody in Immunocytochemistry (ICC/IF)
          LZTFL1 Antibody in Western Blot (WB)
          LZTFL1 Antibody in Flow Cytometry (Flow)

          Product Details

          68281-1-IG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:5,000-1:50,000
          -

          Immunocytochemistry (ICC/IF)

          1:400-1:1,600
          -

          Flow Cytometry (Flow)

          0.4 µg/1x10^6 cell suspension
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          2C1F6

          Immunogen

          Recombinant protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated
          • CoraLite Plus 488
          • Request custom conjugation

          Form

          Liquid

          Purification

          Protein A

          Storage buffer

          PBS with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice

          Product Specific Information

          Immunogen sequence: MAELGLNEHH QNEVINYMRF ARSKRGLRLK TVDSCFQDLK ESRLVEDTFT IDEVSEVLNG LQAVVHSEVE SELINTAYTN VLLLRQLFAQ AEKWYLKLQT DISELENQEL LEQVAEFEKA EITSSNKKPI LDVTKPKLAP LNEGGTAELL NEEILRLQEE NEKLKSRLKT IEIQATNALD EKSKLEKALQ DLQLDQGNQK DFIKAQDLSN LENTVAALKS EFQKTLNDKT ENQKSLEENL ATAKHDLLRV REQLHMAEKE LEKKFQQTAA YRNMKEILTK KNDQIKDLRK RLAQYEPED

          Target Information

          LZTFL1, also known as Leucine Zipper Transcription Factor-Like 1, is situated on the chromosomal region 3p21.3, which is frequently deleted in various cancers, including kidney cancer. It acts as a tumor suppressor by destabilizing AKT, a key protein in cell growth pathways, thus inhibiting tumor cell growth. Additionally, LZTFL1 has been implicated in the epithelial-mesenchymal transition in COVID-19, suggesting a role in viral response pathways. It is associated with Bardet-Biedl syndrome (BBS) and interacts with IFT27, a regulator of sperm flagellum formation, affecting spermatogenesis and male fertility. Knockout models show a phenotype of obesity, retinal degeneration, and abnormal cilia development, highlighting its role in normal ciliary function. The gene has two transcript isoforms and is part of a transcriptional map covering 250 kb, making it a subject of extensive genetic research.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: leucine zipper transcription factor-like 1; Leucine zipper transcription factor-like protein 1

          View more View less

          Gene Aliases: BBS17; LZTFL1

          View more View less

          UniProt ID: (Human) Q9NQ48

          View more View less

          Entrez Gene ID: (Human) 54585

          View more View less

          Function(s)
          protein binding protein complex binding identical protein binding
          Process(es)
          negative regulation of protein localization to cilium negative regulation of protein localization to ciliary membrane
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0