Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Exalpha
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive control: Human brain lysate (Cat. No. X1633C).
Phospholipid-mediated signalling on neurons provokes diverse responses such as neurogenesis, pattern formation and neurite remodelling. A recently uncovered novel set of molecules in the mammalian brain, named plasticity-related genes (PRGs) mediate lipid phosphate phosphatase activity and provide evidence for their involvement in mechanisms of neuronal plasticity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Brain-specific phosphatidic acid phosphatase-like protein 1; Inactive 2-lysophosphatidate phosphatase PLPPR4; Lipid phosphate phosphatase-related protein type 4; Phospholipid phosphatase-related protein type 4; plasticity related gene 1; plasticity related gene-1; Plasticity-related gene 1 protein; PRG-1; RP4-788L13.1
Gene Aliases: KIAA0455; LPPR4; LPR4; PHP1; PLPPR4; PRG-1; PRG1
UniProt ID: (Human) Q7Z2D5
Entrez Gene ID: (Human) 9890
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support