Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Ly-6C Antibodies

          Invitrogen

          Ly-6C Monoclonal Antibody (HK1.4), eFluor™ 450, eBioscience™

          65 Published Figures
          102 References
          View all (18) Ly-6C antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Ly-6C Monoclonal Antibody (HK1.4), eFluor™ 450, eBioscience™

          • Antibody Testing Data (1)
          • Published Figures (65)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Ly-6C Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.

          FIGURE: 1 / 66

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Ly-6C Antibody (48-5932-82) in Flow

          Staining of C57Bl/6 splenocytes with Anti-Mouse CD8a FITC (Product # 11-0081-82) and 0.125 µg of Anti-Mouse Ly-6C eFluor® 450. Total viable cells were used for analysis. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)
          Ly-6C Antibody in Flow Cytometry (Flow)

          Product Details

          48-5932-82

          Applications
          Tested Dilution
          Publications

          Flow Cytometry (Flow)

          0.25 µg/test
          View 102 publications 102 publications
          Product Specifications

          Species Reactivity

          Mouse

          Published species

          Mouse

          Host/Isotype

          Rat / IgG2c, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          HK1.4

          Conjugate

          eFluor™ 450 eFluor™ 450 eFluor™ 450
          • Alexa Fluor 488 
          • APC 
          • APC-eFluor 780
          • Functional Grade
          • PE 
          • PE-Cyanine7
          • PerCP-Cyanine5.5
          • Super Bright 436
          • Request custom conjugation

          Excitation/Emission Max

          405/445 nm View spectra spectra

          Form

          Liquid

          Concentration

          0.2 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.09% sodium azide

          Storage conditions

          4°C, store in dark, DO NOT FREEZE!

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10805519

          Product Specific Information

          Description: The monoclonal antibody HK1.4 recognizes mouse Ly-6C, a GPI-linked protein of the Ly6 family. Ly-6C is found on monocytes/macrophages, endothelial cells and granulocytes as well as a subset of lymphocytes. Some variation of expression is found on different mouse strains in regards to expression on CD4 and CD8 lymphocytes. These correlate to 2 alleles both of which are recognized by HK1.4: Ly6c.1 found on C57Bl/6 and SJL cells which results in staining of both CD4 and CD8 cells while Ly6-C.2 found on BALB/c and 3H/He results in staining of CD8, but not CD4 cells.

          In vitro addition with HK1.4 antibody can increase proliferation and stimulate cytokine release.

          Applications Reported: This HK1.4 antibody has been reported for use in flow cytometric analysis.

          Applications Tested: This HK1.4 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.

          Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          How to use the Panel Builder

          Watch the video to learn how to use the Invitrogen Flow Cytometry Panel Builder to build your next flow cytometry panel in 5 easy steps.

          Bioinformatics

          Protein Aliases: Ly-6C1; Ly6c2; lymphocyte antigen 6 complex, locus C; Lymphocyte antigen 6C1; Lymphocyte antigen Ly-6C

          View more View less

          Gene Aliases: AA682074; AA959465; Ly-6C; Ly-6C1; Ly6c; Ly6c1

          View more View less

          UniProt ID: (Mouse) P0CW02

          View more View less

          Entrez Gene ID: (Mouse) 17067

          View more View less

          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0