Product References
T cells mediate autoantibody-induced cutaneous inflammation and blistering in epidermolysis bullosa acquisita.
Scientific reports
Bieber K,Witte M,Sun S,Hundt JE,Kalies K,Dräger S,Kasprick A,Twelkmeyer T,Manz RA,König P,Köhl J,Zillikens D,Ludwig RJ
MA1-82826 was used in immunohistochemistry - frozen section to study the role of T cells in the effector phase of antibody-mediated autoimmune dermatoses
Mon Dec 05 00:00:00 UTC 2016
Short-chain fatty acids activate GPR41 and GPR43 on intestinal epithelial cells to promote inflammatory responses in mice.
Gastroenterology
Kim MH,Kang SG,Park JH,Yanagisawa M,Kim CH
MA1-82826 was used in immunohistochemistry (paraffin) to investigate the role of short-chain fatty acids and their receptor on intestinal immune responses.
Thu Aug 01 00:00:00 UTC 2013
Consequences of the combined loss of BOK and BAK or BOK and BAX.
Cell death & disease
Ke F,Bouillet P,Kaufmann T,Strasser A,Kerr J,Voss AK
MA1-82826 was used in flow cytometry to determine if Bok and Bax have any functional redundancy controlling apoptosis using knock out mice.
Thu Jun 06 00:00:00 UTC 2013
IL-1α/IL-1R1 expression in chronic obstructive pulmonary disease and mechanistic relevance to smoke-induced neutrophilia in mice.
PloS one
Botelho FM,Bauer CM,Finch D,Nikota JK,Zavitz CC,Kelly A,Lambert KN,Piper S,Foster ML,Goldring JJ,Wedzicha JA,Bassett J,Bramson J,Iwakura Y,Sleeman M,Kolbeck R,Coyle AJ,Humbles AA,Stämpfli MR
MA1-82826 was used in flow cytometry to review cellular and molecular mechanisms that contribute to chronic obstructive pulmonary disease pathogenesis
Fri Jul 20 00:00:00 UTC 2012
Gfi1 integrates progenitor versus granulocytic transcriptional programming.
Blood
Horman SR,Velu CS,Chaubey A,Bourdeau T,Zhu J,Paul WE,Gebelein B,Grimes HL
MA1-82826 was used in flow cytometry to report that Gfi1 suppresses the HoxA9-Pbx1-Meis1 progenitor program and induces a granulopoietic transcription program
Thu May 28 00:00:00 UTC 2009
Pathogenic role of B cells and antibodies in murine Leishmania amazonensis infection.
International journal for parasitology
Wanasen N,Xin L,Soong L
MA1-82826 was used in immunohistochemistry to assess whether B cells and antibodies are involved in Leishmania amazonensis disease and pathogenesis in mice
Sat Mar 01 00:00:00 UTC 2008
Murine neutrophils present Class II restricted antigen.
Immunology letters
Culshaw S,Millington OR,Brewer JM,McInnes IB
MA1-82826 was used in flow cytometry to study neutrophil-T cell interactions
Sun Jun 15 00:00:00 UTC 2008
Characterization of early gamma interferon (IFN-gamma) expression during murine listeriosis: identification of NK1.1+ CD11c+ cells as the primary IFN-gamma-expressing cells.
Infection and immunity
Chang SR,Wang KJ,Lu YF,Yang LJ,Chen WJ,Lin YH,Chang HH,Wang SL
MA1-82826 was used in flow cytometry to identify the cells that are producing gamma interferon during infection with Listeria monocytogenes
Thu Mar 01 00:00:00 UTC 2007
Hematopoiesis and stem cell renewal in long-term bone marrow cultures containing catalase.
Blood
Gupta R,Karpatkin S,Basch RS
MA1-82826 was used in flow cytometry to investigate how peroxide affects hematopoiesis
Wed Mar 01 00:00:00 UTC 2006
Hydrolytic and nonenzymatic functions of acetylcholinesterase comodulate hemopoietic stress responses.
Journal of immunology (Baltimore, Md. : 1950)
Grisaru D,Pick M,Perry C,Sklan EH,Almog R,Goldberg I,Naparstek E,Lessing JB,Soreq H,Deutsch V
MA1-82826 was used in flow cytometry to demonstrate that AChE-R and ARP facilitate granulocytosis
Sun Jan 01 00:00:00 UTC 2006
Expression of chitinase-like proteins in the skin of chronic proliferative dermatitis (cpdm/cpdm) mice.
Experimental dermatology
HogenEsch H,Dunham A,Seymour R,Renninger M,Sundberg JP
MA1-82826 was used in immunohistochemistry - frozen section to explore the role of mammalian chitinase-like proteins in dermatitis
Sun Oct 01 00:00:00 UTC 2006
The earliest thymic progenitors in adults are restricted to T, NK, and dendritic cell lineage and have a potential to form more diverse TCRbeta chains than fetal progenitors.
Journal of immunology (Baltimore, Md. : 1950)
Lu M,Tayu R,Ikawa T,Masuda K,Matsumoto I,Mugishima H,Kawamoto H,Katsura Y
MA1-82826 was used in flow cytometry to study T cell progenitors in the adult thymus
Tue Nov 01 00:00:00 UTC 2005
Effect of thermal injury on splenic myelopoiesis.
Shock (Augusta, Ga.)
Noel JG,Guo X,Wells-Byrum D,Schwemberger S,Caldwell CC,Ogle CK
MA1-82826 was used in flow cytometry to measure the number of splenic myeloid progenitors and their progeny after burn injury
Tue Feb 01 00:00:00 UTC 2005
In vivo self-renewing divisions of haematopoietic stem cells are increased in the absence of the early G1-phase inhibitor, p18INK4C.
Nature cell biology
Yuan Y,Shen H,Franklin DS,Scadden DT,Cheng T
MA1-82826 was used in flow cytometry to demonstrate that deletion p18(INK4C) results in improved long-term engraftment
Sat May 01 00:00:00 UTC 2004
Chemotaxis inhibitory protein of Staphylococcus aureus, a bacterial antiinflammatory agent.
The Journal of experimental medicine
de Haas CJ,Veldkamp KE,Peschel A,Weerkamp F,Van Wamel WJ,Heezius EC,Poppelier MJ,Van Kessel KP,van Strijp JA
MA1-82826 was used in flow cytometry to identify and characterize the chemotaxis inhibitory protein of Staphylococcus aureus
Mon Mar 01 00:00:00 UTC 2004
Schistosoma mansoni worms induce anergy of T cells via selective up-regulation of programmed death ligand 1 on macrophages.
Journal of immunology (Baltimore, Md. : 1950)
Smith P,Walsh CM,Mangan NE,Fallon RE,Sayers JR,McKenzie AN,Fallon PG
MA1-82826 was used in flow cytometry to elucidate how Schistosoma mansoni suppresses T cell activation
Thu Jul 15 00:00:00 UTC 2004
Regulation of thymus size by competition for stromal niches among early T cell progenitors.
Journal of immunology (Baltimore, Md. : 1950)
Prockop SE,Petrie HT
MA1-82826 was used in flow cytometry to investigate mechanisms that regulate thymus size
Sun Aug 01 00:00:00 UTC 2004
Passive transfer of flt-3L-derived dendritic cells delays diabetes development in NOD mice and associates with early production of interleukin (IL)-4 and IL-10 in the spleen of recipient mice.
Clinical and experimental immunology
Morin J,Faideau B,Gagnerault MC,Lepault F,Boitard C,Boudaly S
MA1-82826 was used in flow cytometry to assess the contribution of DC function in non-obese diabetic mice.
Mon Dec 01 00:00:00 UTC 2003
Host conditioning with total lymphoid irradiation and antithymocyte globulin prevents graft-versus-host disease: the role of CD1-reactive natural killer T cells.
Biology of blood and marrow transplantation : journal of the American Society for Blood and Marrow Transplantation
Lan F,Zeng D,Higuchi M,Higgins JP,Strober S
MA1-82826 was used in flow cytometry to test if a total lymphoid irradiation and anti-thymocyte serum regimen can prevent graft verses host disease.
Sun Jun 01 00:00:00 UTC 2003
Granulocyte-dendritic cell unbalance in the non-obese diabetic mice.
Cellular immunology
Morin J,Chimènes A,Boitard C,Berthier R,Boudaly S
MA1-82826 was used in flow cytometry to investigate DCs in non-obese diabetic mice.
Thu May 01 00:00:00 UTC 2003
The immunogenicity, protective efficacy and safety of BBG2Na, a subunit respiratory syncytial virus (RSV) vaccine candidate, against RSV-B.
Vaccine
Power UF,Plotnicky H,Blaecke A,Nguyen TN
MA1-82826 was used in flow cytometry to determine the safety of using BBG2Na in animals challenged with RSV-B.
Fri Dec 12 00:00:00 UTC 2003
Enhanced pulmonary immunopathology following neonatal priming with formalin-inactivated respiratory syncytial virus but not with the BBG2NA vaccine candidate.
Vaccine
Plotnicky H,Siegrist CA,Aubry JP,Bonnefoy JY,Corvaïa N,Nguyen TN,Power UF
MA1-82826 was used in flow cytometry to study the effect of neonatal priming on the Th2 response using respiratory syncytial virus.
Mon Jun 02 00:00:00 UTC 2003
Predominance of NK1.1+TCR alpha beta+ or DX5+TCR alpha beta+ T cells in mice conditioned with fractionated lymphoid irradiation protects against graft-versus-host disease: "natural suppressor" cells.
Journal of immunology (Baltimore, Md. : 1950)
Lan F,Zeng D,Higuchi M,Huie P,Higgins JP,Strober S
MA1-82826 was used in flow cytometry to develop a nonmyeloablative host conditioning regimen for transplantation.
Wed Aug 15 00:00:00 UTC 2001
Tumor-infiltrating macrophages induce apoptosis in activated CD8(+) T cells by a mechanism requiring cell contact and mediated by both the cell-associated form of TNF and nitric oxide.
Journal of immunology (Baltimore, Md. : 1950)
Saio M,Radoja S,Marino M,Frey AB
MA1-82826 was used in flow cytometry to examine the ability of different tumor cells to induce apoptosis of activated CD8(+) T cells in vitro.
Thu Nov 15 00:00:00 UTC 2001
Lipophilic quaternary ammonium salt acts as a mucosal adjuvant when co-administered by the nasal route with vaccine antigens.
Vaccine
Klinguer C,Beck A,De-Lys P,Bussat MC,Blaecke A,Derouet F,Bonnefoy JY,Nguyen TN,Corvaïa N,Velin D
MA1-82826 was used in immunohistochemistry to characterize a new nasal adjuvant, dimethyldioctadecylammonium bromide.
Fri Jul 20 00:00:00 UTC 2001
BBG2Na an RSV subunit vaccine candidate intramuscularly injected to human confers protection against viral challenge after nasal immunization in mice.
Vaccine
Goetsch L,Plotnicky-Gilquin H,Aubry JP,De-Lys P,Haeuw JF,Bonnefoy JY,Nguyen NT,Corvaïa N,Velin D
MA1-82826 was used in immunohistochemistry to discuss vaccination with BBG2Na.
Mon Jul 16 00:00:00 UTC 2001
Reduced brain edema after traumatic brain injury in mice deficient in P-selectin and intercellular adhesion molecule-1.
Journal of leukocyte biology
Whalen MJ,Carlos TM,Dixon CE,Robichaud P,Clark RS,Marion DW,Kochanek PM
MA1-82826 was used in immunohistochemistry (frozen) to study the contributions of P-selectin and ICAM-1 to traumatic brain injury.
Tue Feb 01 00:00:00 UTC 2000
Polymorphonuclear neutrophils are necessary for the recruitment of CD8(+) T cells in the liver in a pregnant mouse model of Chlamydophila abortus (Chlamydia psittaci serotype 1) infection.
Infection and immunity
de Oca RM,Buendía AJ,Del Río L,Sánchez J,Salinas J,Navarro JA
MA1-82826 was used in immunohistochemistry (frozen) to determine the role of polymorphonuclear neutrophils in Chlamydophila abortus-infected mice.
Wed Mar 01 00:00:00 UTC 2000
Comparison of erythropoietic response to androgen in young and old senescence accelerated mice.
Mechanisms of ageing and development
Saitoh T,Morimoto K,Kumagai T,Tsuboi I,Aikawa S,Horie T
MA1-82826 was used in flow cytometry to test if the functional capacity of hemopoietic progenitor cells and the micro-environment of aged and young mice are identical.
Mon Aug 30 00:00:00 UTC 1999
Unimpaired macrophage differentiation and activation in mice lacking the zinc finger transplantation factor NGFI-A (EGR1).
Molecular and cellular biology
Lee SL,Wang Y,Milbrandt J
MA1-82826 was used in flow cytometry to examine the effect of NGFI-A deficiency on macrophage differentiation and function in vivo.
Thu Aug 01 00:00:00 UTC 1996
Leukocyte-endothelial cell recognition: evidence of a common molecular mechanism shared by neutrophils, lymphocytes, and other leukocytes.
Journal of immunology (Baltimore, Md. : 1950)
Lewinsohn DM,Bargatze RF,Butcher EC
MA1-82826 was used in flow cytometry to show the involvement of the MEL-14 antigen in neutrophil-endothelial cell interactions
Mon Jun 15 00:00:00 UTC 1987