Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • MAGED1 Antibodies

          Abnova

          MAGED1 Polyclonal Antibody, MaxPab™

          View all (12) MAGED1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite MAGED1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          MAGED1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MAGED1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MAGED1 Antibody (H00009500-B01P) in WB

          Western Blot analysis of MAGED1 expression in transfected 293T cell line by MAGED1 MaxPab polyclonal antibody. Lane 1: MAGED1 transfected lysate(85.58 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MAGED1 Antibody in Western Blot (WB)

          Product Details

          H00009500-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MAGED1 (NP_001005332.1, 1 a.a. approximately 778 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAQKMDCGAG LLGFQAEASV EDSALLMQTL MEAIQISEAP PTNQATAAAS PQSSQPPTAN EMADIQVSAA AARPKSAFKV QNATTKGPNG VYDFSQAHNA KDVPNTQPKA AFKSQNATPK GPNAAYDFSQ AATTGELAAN KSEMAFKAQN ATTKVGPNAT YNFSQSLNAN DLANSRPKTP FKAWNDTTKA PTADTQTQNV NQAKMATSQA DIETDPGISE PDGATAQTSA DGSQAQNLES RTIIRGKRTR KINNLNVEEN SSGDQRRAPL AAGTWRSAPV PVTTQNPPGA PPNVLWQTPL AWQNPSGWQN QTARQTPPAR QSPPARQTPP AWQNPVAWQN PVIWPNPVIW QNPVIWPNPI VWPGPVVWPN PLAWQNPPGW QTPPGWQTPP GWQGPPDWQG PPDWPLPPDW PLPPDWPLPT DWPLPPDWIP ADWPIPPDWQ NLRPSPNLRP SPNSRASQNP GAAQPRDVAL LQERANKLVK YLMLKDYTKV PIKRSEMLRD IIREYTDVYP EIIERACFVL EKKFGIQLKE IDKEEHLYIL ISTPESLAGI LGTTKDTPKL GLLLVILGVI FMNGNRASEA VLWEALRKMG LRPGVRHPLL GDLRKLLTYE FVKQKYLDYR RVPNSNPPEY EFLWGLRSYH ETSKMKVLRF IAEVQKRDPR DWTAQFMEAA DEALDALDAA AAEAEARAEA RTRMGIGDEA VSGPWSWDDI EFELLTWDEE GDFGDPWSRI PFTFWARYHQ NARSRFPQTF AGPIIGPGGT ASANFAANFG AIGFFWVE

          Target Information

          This gene is a member of the melanoma antigen gene (MAGE) family. Most of the genes of this family encode tumor specific antigens that are not expressed in normal adult tissues except testis. Although the protein encoded by this gene shares strong homology with members of the MAGE family, it is expressed in almost all normal adult tissues. This gene has been demonstrated to be involved in the p75 neurotrophin receptor mediated programmed cell death pathway. Three transcript variants encoding two different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 11B6; BCG-1; JCL-1; MAGE tumor antigen CCF; MAGE-D1 antigen; Maged2; melanoma antigen family D, 1; melanoma antigen family D1; Melanoma-associated antigen D1; Neurotrophin receptor-interacting MAGE homolog

          View more View less

          Gene Aliases: DLXIN-1; MAGED1; NRAGE; PP2250; PRO2292

          View more View less

          UniProt ID: (Human) Q5VSH6

          View more View less

          Entrez Gene ID: (Human) 9500

          View more View less

          Function(s)
          transcription coactivator activity protein binding
          Process(es)
          regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter circadian regulation of gene expression regulation of circadian rhythm regulation of apoptotic process positive regulation of MAP kinase activity negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated negative regulation of epithelial cell proliferation positive regulation of branching involved in ureteric bud morphogenesis positive regulation of apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-48zfn:80/100.66.135.186:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0